Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006660.4 | NP_006651.2 | 8 | 1249 | ||
NR_133680.1 | 8 | 1137 | ||||
XM_011521164.2 | XP_011519466.1 | 7 | 1191 | |||
GenBank mRNA | AJ006267.3 | - | 8 | 1054 | ||
AK292316.1 | - | 8 | 1175 | |||
AK308133.1 | - | 8 | 1169 | |||
AK310614.1 | - | 8 | 1171 | |||
AL136922.1 | - | 8 | 1152 | |||
BC130373.1 | - | 8 | 1045 | 40146404 | ||
BC136487.1 | - | 8 | 1045 | 9020476 |