Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130690.2 | NP_001124162.1 | 12 | 1147 | ||
NM_006661.3 | NP_006652.1 | 13 | 1275 | |||
XM_006715321.3 | XP_006715384.1 | 12 | 2472 | |||
XM_011535387.2 | XP_011533689.2 | 14 | 3267 | |||
XM_011535388.2 | XP_011533690.1 | 13 | 2600 | |||
XM_011535393.2 | XP_011533695.1 | 12 | 1075 | |||
XM_017010194.1 | XP_016865683.1 | 14 | 3267 | |||
XM_017010195.1 | XP_016865684.1 | 12 | 1020 | |||
XM_017010196.1 | XP_016865685.1 | 14 | 2718 | |||
XM_017010197.1 | XP_016865686.1 | 14 | 3267 | |||
GenBank mRNA | AB020593.1 | - | 13 | 1280 | ||
AB026816.1 | - | 12 | 1080 | |||
AF127479.1 | - | 12 | 1096 | |||
AF127480.1 | - | 12 | 1116 | |||
AK297555.1 | - | 3 | 226 | |||
BC104858.1 | - | 12 | 1067 | 8143861 | ||
BC104860.1 | - | 12 | 1067 | 8143863 | ||
BC143297.1 | - | 13 | 1186 | |||
BC143301.1 | - | 13 | 1186 | |||
CR536567.1 | - | 12 | 1030 | |||
KU232567.1 | - | 12 | 1518 |