Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130690.2 | NP_001124162.1 | 7, 8 | 784 | ||
NM_006661.3 | NP_006652.1 | 8, 9 | 912 | |||
XM_006715321.3 | XP_006715384.1 | 7, 8 | 2109 | |||
XM_011535387.2 | XP_011533689.2 | 9, 10 | 2904 | |||
XM_011535388.2 | XP_011533690.1 | 8, 9 | 2237 | |||
XM_011535393.2 | XP_011533695.1 | 6, 7 | 557 | |||
XM_017010194.1 | XP_016865683.1 | 9, 10 | 2904 | |||
XM_017010195.1 | XP_016865684.1 | 7, 8 | 657 | |||
XM_017010196.1 | XP_016865685.1 | 9, 10 | 2355 | |||
XM_017010197.1 | XP_016865686.1 | 9, 10 | 2904 | |||
GenBank mRNA | AB020593.1 | - | 8, 9 | 917 | ||
AB026816.1 | - | 7, 8 | 717 | |||
AF127479.1 | - | 7, 8 | 733 | |||
AF127480.1 | - | 7, 8 | 753 | |||
BC104858.1 | - | 7, 8 | 704 | 8143861 | ||
BC104860.1 | - | 7, 8 | 704 | 8143863 | ||
BC143297.1 | - | 8, 9 | 823 | |||
BC143301.1 | - | 8, 9 | 823 | |||
CR536567.1 | - | 7, 8 | 667 | |||
KU232567.1 | - | 7, 8 | 1155 |