Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006662.2 | NP_006653.2 | 21 | 3896 | ||
GenBank mRNA | AB621816.1 | - | 21 | 3963 | ||
AF143946.1 | - | 17 | 3130 | |||
AK023808.1 | - | 14 | 2606 | |||
AK128030.1 | - | 21 | 3896 | |||
DA109400.1 | - | 1 | 202 |