Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R13L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142502.1 | NP_001135974.1 | 10 | 2098 | ||
NM_006663.3 | NP_006654.2 | 10 | 2096 | |||
XM_017026177.1 | XP_016881666.1 | 11 | 2154 | |||
XM_017026178.1 | XP_016881667.1 | 11 | 2214 | |||
GenBank mRNA | AF078036.1 | - | 4 | 1077 | ||
AF078037.1 | - | 3 | 1531 | |||
AJ888472.1 | - | 9 | 2019 | |||
AY869712.1 | - | 10 | 2077 | |||
BC001475.2 | - | 4 | 683 | |||
BC020589.1 | - | 3 | 337 | |||
BC032298.1 | - | 10 | 2085 | 4994121 | ||
BC064913.1 | - | 10 | 2061 | 6068949 | ||
DQ986418.1 | - | 3 | 756 |