Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008563.4 | NP_001008563.2 | 17 | 2017 | ||
NM_001110303.3 | NP_001103773.2 | 17 | 1919 | |||
NM_006676.7 | NP_006667.3 | 17 | 1919 | |||
XM_005251665.3 | XP_005251722.1 | 17 | 1907 | |||
XM_005251666.3 | XP_005251723.1 | 12 | 1481 | |||
XM_011518161.2 | XP_011516463.1 | 18 | 2259 | |||
XM_011518162.2 | XP_011516464.1 | 17 | 2438 | |||
XM_017014221.1 | XP_016869710.1 | 12 | 1481 | |||
GenBank mRNA | AB023220.1 | - | 16 | 1837 | ||
AY074877.1 | - | 17 | 1953 | |||
BC039593.1 | - | 17 | 1863 | 5588842 |