Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ME3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014811.1 | NP_001014811.1 | 9 | 1355 | ||
NM_001161586.1 | NP_001155058.1 | 10 | 1428 | |||
NM_006680.2 | NP_006671.2 | 10 | 1203 | |||
XM_005273717.1 | XP_005273774.1 | 10 | 1304 | |||
XM_017017136.1 | XP_016872625.1 | 10 | 2186 | |||
XM_017017137.1 | XP_016872626.1 | 6 | 728 | |||
XM_017017138.1 | XP_016872627.1 | 11 | 1312 | |||
GenBank mRNA | AA403046.1 | - | 5 | 443 | ||
AK313695.1 | - | 10 | 1278 | |||
BC022472.2 | - | 9 | 1355 | 4812045 | ||
BQ882896.1 | - | 1 | 88 | |||
X79440.1 | - | 9 | 1133 |