Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016540.3 | NP_057624.3 | 4 | 1228 | ||
XM_011542559.2 | XP_011540861.1 | 3 | 1162 | |||
GenBank mRNA | AB040973.1 | - | 4 | 1061 | ||
AF236081.1 | - | 4 | 1056 | |||
AK315161.1 | - | 4 | 1228 | |||
BC067473.1 | - | 4 | 1056 | 6971838 | ||
BC067474.1 | - | 4 | 1056 | 6971839 | ||
EU432120.1 | - | 4 | 1056 |
Set Membership: |
G-protein coupled receptor |