Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016131.4 | NP_057215.3 | 1 | 849 | ||
GenBank mRNA | AF086917.1 | - | 1 | 194 | ||
AF091032.1 | - | 1 | 40 | |||
AF106681.1 | - | 1 | 190 | |||
AF297660.1 | - | 1 | 100 | |||
AF498945.1 | - | 1 | 100 | |||
AK023223.1 | - | 1 | 599 | |||
AL136650.1 | - | 1 | 608 | |||
AL517754.3 | - | 1 | 610 | |||
BC000896.1 | - | 1 | 175 | 3464547 | ||
CR457303.1 | - | 1 | 100 |