Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006690.3 | NP_006681.1 | 2, 3 | 392 | ||
XM_011528500.2 | XP_011526802.1 | 2, 3 | 389 | |||
XM_017027597.1 | XP_016883086.1 | 2, 3 | 389 | |||
XM_017027598.1 | XP_016883087.1 | 2, 3 | 363 | |||
GenBank mRNA | AB021227.1 | - | 2, 3 | 392 | ||
AF131284.1 | - | 2, 3 | 389 | |||
BC047614.1 | - | 2, 3 | 332 | |||
DQ300264.1 | - | 3, 4 | 233 |