Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006699.3 | NP_006690.1 | 1 | 814 | ||
XM_006710302.2 | XP_006710365.1 | 1 | 822 | |||
XM_011540536.2 | XP_011538838.1 | 1 | 884 | |||
XM_017000115.1 | XP_016855604.1 | 1 | 884 | |||
GenBank mRNA | AF027156.1 | - | 1 | 613 | ||
AK307842.1 | - | 1 | 822 | |||
AU133800.1 | - | 1 | 334 | |||
BC063300.1 | - | 1 | 812 | 5263292 |