Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318060.1 | NP_001304989.1 | 11 | 2174 | ||
NM_001318061.1 | NP_001304990.1 | 12 | 2189 | |||
NM_001318062.1 | NP_001304991.1 | 12 | 2220 | |||
NM_006716.3 | NP_006707.1 | 12 | 2281 | |||
GenBank mRNA | AB028069.1 | - | 12 | 2294 | ||
AB028070.1 | - | 12 | 2233 | |||
AF160249.1 | - | 12 | 1993 | |||
AF160876.1 | - | 12 | 1976 | |||
AK022969.1 | - | 1 | 641 | |||
AK292569.1 | - | 12 | 2023 | |||
AK303175.1 | - | 12 | 1885 | |||
BC036045.1 | - | 12 | 2087 | 5273059 | ||
BC047693.1 | - | 12 | 1962 | 5753567 |