Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145860.1 | NP_001139332.1 | 13 | 1880 | ||
NM_001145861.1 | NP_001139333.1 | 13 | 1820 | |||
NM_015029.2 | NP_055844.2 | 13 | 1808 | |||
GenBank mRNA | AK000837.1 | - | 7 | 874 | ||
AK291434.1 | - | 13 | 1877 | |||
BC011529.1 | - | 13 | 1808 | 3853321 | ||
D31765.1 | - | 10 | 1419 |