Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006825.3 | NP_006816.2 | 2 | 1842 | ||
GenBank mRNA | AK123693.1 | - | 3 | 1672 | ||
AK223563.1 | - | 2 | 1896 | |||
BC025341.1 | - | 3 | 1633 | |||
BC067357.1 | - | 3 | 1752 | |||
BC082972.1 | - | 2 | 1815 | 6084968 | ||
BC094824.1 | - | 3 | 1623 | 6374931 | ||
X69910.2 | - | 2 | 1842 |