Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256420.1 | NP_001243349.1 | 1 | 231 | ||
NM_014268.3 | NP_055083.1 | 1 | 231 | |||
GenBank mRNA | AB016823.1 | - | 1 | 166 | ||
AK090945.1 | - | 1 | 187 | |||
AK315766.1 | - | 1 | 184 | |||
BC007318.2 | - | 1 | 174 | 2989643 | ||
BT020086.1 | - | 1 | 51 | |||
CR536545.1 | - | 1 | 51 | |||
DC354326.1 | - | 1 | 231 | |||
X94232.1 | - | 1 | 191 |