Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006835.2 | NP_006826.1 | 7 | 1413 | ||
XM_011531555.1 | XP_011529857.1 | 5 | 1238 | |||
GenBank mRNA | AB209766.1 | - | 7 | 1415 | ||
AF135162.1 | - | 7 | 1068 | |||
AK222931.1 | - | 6 | 865 | |||
AK301111.1 | - | 6 | 1274 | |||
AK312632.1 | - | 9 | 1283 | |||
BC000420.2 | - | 7 | 1412 | 2964432 | ||
BC004975.1 | - | 7 | 1412 | 2957878 | ||
BT020019.1 | - | 6 | 869 | |||
CR541783.1 | - | 6 | 869 | |||
D50310.1 | - | 6 | 869 |