Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006835.2 | NP_006826.1 | 7 | 1532 | ||
XM_011531555.1 | XP_011529857.1 | 5 | 1357 | |||
GenBank mRNA | AB209766.1 | - | 7 | 1534 | ||
AF135162.1 | - | 7 | 1187 | |||
AK222931.1 | - | 6 | 984 | |||
AK301111.1 | - | 6 | 1393 | |||
AK312632.1 | - | 9 | 1402 | |||
BC000420.2 | - | 7 | 1531 | 2964432 | ||
BC004975.1 | - | 7 | 1531 | 2957878 | ||
BT020019.1 | - | 6 | 988 | |||
CR541783.1 | - | 6 | 988 | |||
D50310.1 | - | 6 | 988 |