Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC27A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321196.1 | NP_001308125.1 | 1 | 455 | ||
NM_012254.2 | NP_036386.1 | 1 | 455 | |||
XM_011526364.1 | XP_011524666.1 | 1 | 384 | |||
XM_017026214.1 | XP_016881703.1 | 1 | 384 | |||
GenBank mRNA | AB208931.1 | - | 1 | 803 | ||
AF064255.1 | - | 1 | 415 | |||
AK123036.1 | - | 1 | 455 | |||
AK298446.1 | - | 1 | 382 |