Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC27A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005094.3 | NP_005085.2 | 3 | 610 | ||
XM_017014222.1 | XP_016869711.1 | 4 | 483 | |||
GenBank mRNA | AF055899.1 | - | 2 | 325 | ||
AK027504.1 | - | 2 | 190 | |||
AK290222.1 | - | 3 | 610 | |||
AK314681.1 | - | 3 | 450 | |||
BC065003.1 | - | 3 | 609 | 6023438 |