Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006851.2 | NP_006842.2 | 1 | 259 | ||
XM_011537775.2 | XP_011536077.1 | 1 | 238 | |||
GenBank mRNA | AF400440.1 | - | 1 | 139 | ||
AK304847.1 | - | 1 | 239 | |||
AK313449.1 | - | 1 | 125 | |||
BC012510.1 | - | 1 | 175 | 4453841 | ||
CB123261.1 | - | 1 | 312 | |||
EF667354.1 | - | 1 | 111 | |||
U16307.3 | - | 1 | 195 | |||
X91911.1 | - | 1 | 239 |