Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBPMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008710.2 | NP_001008710.1 | 3 | 755 | ||
NM_001008711.2 | NP_001008711.1 | 3 | 755 | |||
NM_001008712.2 | NP_001008712.1 | 3 | 755 | |||
NM_006867.3 | NP_006858.1 | 3 | 755 | |||
XM_017012980.1 | XP_016868469.1 | 3 | 750 | |||
XM_017012981.1 | XP_016868470.1 | 3 | 750 | |||
XM_017012982.1 | XP_016868471.1 | 3 | 750 | |||
XM_017012983.1 | XP_016868472.1 | 3 | 1736 | |||
XM_017012984.1 | XP_016868473.1 | 3 | 750 | |||
XM_017012986.1 | XP_016868475.1 | 3 | 750 | |||
XM_017012987.1 | XP_016868476.1 | 3 | 750 | |||
XM_017012988.1 | XP_016868477.1 | 3 | 750 | |||
XM_017012989.1 | XP_016868478.1 | 3 | 750 | |||
XM_017012990.1 | XP_016868479.1 | 3 | 750 | |||
XM_017012991.1 | XP_016868480.1 | 3 | 843 | |||
XM_017012992.1 | XP_016868481.1 | 3 | 750 | |||
GenBank mRNA | AK304858.1 | - | 4 | 573 | ||
AK310533.1 | - | 3 | 183 | |||
BC003608.2 | - | 3 | 696 | 3505124 | ||
BC063494.1 | - | 3 | 162 | |||
BC092476.1 | - | 3 | 243 | 4837343 | ||
D84107.1 | - | 3 | 729 | |||
D84108.1 | - | 3 | 729 | |||
D84109.1 | - | 3 | 729 | |||
D84110.1 | - | 3 | 729 | |||
D84111.1 | - | 1 | 29 |