Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284308.1 | NP_001271237.1 | 5 | 487 | ||
NM_001284309.1 | NP_001271238.1 | 5 | 427 | |||
NM_001284310.1 | NP_001271239.1 | 4 | 402 | |||
NM_001284311.1 | NP_001271240.1 | 3 | 255 | |||
NM_006869.3 | NP_006860.1 | 5 | 660 | |||
GenBank mRNA | AB097049.1 | - | 5 | 634 | ||
AF082324.1 | - | 5 | 634 | |||
AJ006422.1 | - | 5 | 656 | |||
AK092471.1 | - | 4 | 404 | |||
AK124979.1 | - | 3 | 506 | |||
AK290517.1 | - | 6 | 606 | |||
AK300644.1 | - | 3 | 239 | |||
AK300859.1 | - | 3 | 242 | |||
AK300999.1 | - | 5 | 487 | |||
BC033747.1 | - | 5 | 461 | 5195416 | ||
BM837987.1 | - | 3 | 255 | |||
DA872042.1 | - | 5 | 427 |