Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284308.1 | NP_001271237.1 | 6 | 561 | ||
NM_001284309.1 | NP_001271238.1 | 6 | 501 | |||
NM_001284310.1 | NP_001271239.1 | 5 | 476 | |||
NM_001284311.1 | NP_001271240.1 | 4 | 329 | |||
NM_006869.3 | NP_006860.1 | 6 | 734 | |||
GenBank mRNA | AB097049.1 | - | 6 | 708 | ||
AF082324.1 | - | 6 | 708 | |||
AJ006422.1 | - | 6 | 730 | |||
AK092471.1 | - | 5 | 478 | |||
AK124979.1 | - | 4 | 580 | |||
AK290517.1 | - | 7 | 680 | |||
AK300859.1 | - | 4 | 316 | |||
AK300999.1 | - | 6 | 561 | |||
BC033747.1 | - | 6 | 535 | 5195416 | ||
BM837987.1 | - | 4 | 329 | |||
DA872042.1 | - | 6 | 501 |