Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPK1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281443.1 | NP_001268372.1 | 9 | 870 | ||
NM_007000.3 | NP_008931.1 | 8 | 772 | |||
GenBank mRNA | AF085807.1 | - | 7 | 740 | ||
BC107097.1 | - | 8 | 843 | 40015698 | ||
BC107098.1 | - | 7 | 745 | 40015700 |