Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007006.2 | NP_008937.1 | 2, 3 | 468 | ||
GenBank mRNA | AJ001810.1 | - | 2, 3 | 321 | ||
AK312721.1 | - | 2, 3 | 471 | |||
AL535307.3 | - | 2, 3 | 468 | |||
BC001403.2 | - | 2, 3 | 429 | 3139379 | ||
CR456919.1 | - | 2, 3 | 321 |