Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007011.7 | NP_008942.3 | 10 | 1592 | ||
NM_152924.4 | NP_690888.1 | 6 | 1194 | |||
XM_005254832.1 | XP_005254889.1 | 9 | 1230 | |||
XM_005254833.1 | XP_005254890.1 | 6 | 838 | |||
XM_011521165.1 | XP_011519467.1 | 7 | 1043 | |||
GenBank mRNA | AF546700.1 | - | 6 | 926 | ||
AK222841.1 | - | 6 | 866 | |||
AK223083.1 | - | 6 | 864 | |||
AK290340.1 | - | 6 | 866 | |||
AK296827.1 | - | 4 | 449 | |||
BC019248.1 | - | 10 | 1322 | 4826878 | ||
BC090052.1 | - | 6 | 928 | 30388486 | ||
X12433.1 | - | 6 | 849 |