Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007013.3 | NP_008944.1 | 17 | 2056 | ||
XM_005250760.3 | XP_005250817.1 | 16 | 2017 | |||
XM_005250761.3 | XP_005250818.1 | 17 | 2097 | |||
XM_011516802.2 | XP_011515104.1 | 15 | 1745 | |||
XM_017012993.1 | XP_016868482.1 | 16 | 2009 | |||
XM_017012994.1 | XP_016868483.1 | 15 | 1935 | |||
XM_017012995.1 | XP_016868484.1 | 15 | 1878 | |||
XM_017012996.1 | XP_016868485.1 | 14 | 1796 | |||
XM_017012997.1 | XP_016868486.1 | 13 | 1703 | |||
GenBank mRNA | AK292800.1 | - | 17 | 2071 | ||
AK310683.1 | - | 13 | 1430 | |||
AL136739.1 | - | 15 | 1786 | |||
AY043361.1 | - | 16 | 1861 | |||
AY345857.1 | - | 17 | 2090 | |||
BC036065.1 | - | 17 | 2060 | 5296005 | ||
U96113.1 | - | 10 | 1210 |