Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB561D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291284.1 | NP_001278213.1 | 4 | 639 | ||
NM_007022.4 | NP_008953.1 | 4 | 705 | |||
GenBank mRNA | AF040704.1 | - | 4 | 462 | ||
AK291167.1 | - | 4 | 525 | |||
BC047691.1 | - | 4 | 534 | 6060648 | ||
BX641103.1 | - | 1 | 2546 |