Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007027.3 | NP_008958.2 | 13 | 2269 | ||
XM_005247076.3 | XP_005247133.1 | 13 | 2741 | |||
XM_011512357.2 | XP_011510659.1 | 13 | 2340 | |||
XM_017005636.1 | XP_016861125.1 | 13 | 2339 | |||
XM_017005637.1 | XP_016861126.1 | 10 | 1793 | |||
GenBank mRNA | AB019397.1 | - | 13 | 2216 | ||
AK302584.1 | - | 13 | 2235 | |||
BC126209.1 | - | 12 | 2159 | 8991925 | ||
BC151237.1 | - | 13 | 2222 | 8860504 | ||
D87448.1 | - | 13 | 2222 |