Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007033.4 | NP_008964.3 | 4 | 451 | ||
XM_005244713.4 | XP_005244770.2 | 4 | 1272 | |||
XM_006710306.2 | XP_006710369.1 | 5 | 533 | |||
XM_011540542.1 | XP_011538844.1 | 5 | 471 | |||
XM_011540543.1 | XP_011538845.1 | 4 | 319 | |||
XM_017000131.1 | XP_016855620.1 | 4 | 1802 | |||
XM_017000132.1 | XP_016855621.1 | 5 | 561 | |||
XM_017000133.1 | XP_016855622.1 | 5 | 644 | |||
XM_017000134.1 | XP_016855623.1 | 6 | 558 | |||
XM_017000135.1 | XP_016855624.1 | 4 | 558 | |||
GenBank mRNA | AF091071.1 | - | 4 | 336 | ||
AF157324.1 | - | 4 | 390 | |||
AJ001421.1 | - | 3 | 260 | |||
AK095750.1 | - | 3 | 2046 | |||
AK289658.1 | - | 4 | 385 | |||
BC004965.1 | - | 4 | 386 | 3546970 | ||
BT007224.1 | - | 3 | 260 | |||
CR456806.1 | - | 3 | 260 | |||
DA409924.1 | - | 5 | 533 |