Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317099.1 | NP_001304028.1 | 3 | 707 | ||
NM_001317100.1 | NP_001304029.1 | 3 | 901 | |||
NM_001317101.1 | NP_001304030.1 | 2 | 788 | |||
NM_001317102.1 | NP_001304031.1 | 2 | 462 | |||
NM_007034.4 | NP_008965.2 | 2 | 820 | |||
GenBank mRNA | AB209922.1 | - | 2 | 698 | ||
AK297985.1 | - | 2 | 370 | |||
AK313205.1 | - | 2 | 706 | |||
BC034721.1 | - | 2 | 696 | 4340658 | ||
BX441874.2 | - | 2 | 663 | |||
DC349099.1 | - | 3 | 508 | |||
DC412631.1 | - | 2 | 463 | |||
U40992.2 | - | 2 | 713 |