Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021794.3 | NP_068566.2 | 1 | 264 | ||
GenBank mRNA | AF171932.1 | - | 1 | 76 | ||
AF171933.1 | - | 1 | 76 | |||
AK292483.1 | - | 1 | 255 | |||
AY358734.1 | - | 1 | 238 | |||
BC028372.2 | - | 1 | 235 | 4831258 |