Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007038.4 | NP_008969.2 | 1, 2 | 1837 | ||
XM_011529448.2 | XP_011527750.1 | 1, 2 | 1837 | |||
GenBank mRNA | AF142099.1 | - | 1, 2 | 1230 | ||
AK094057.1 | - | 1, 2 | 1836 | |||
BC093775.1 | - | 1, 2 | 1192 | 7939620 | ||
BC093777.1 | - | 1, 2 | 1192 | 7939622 |