Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007038.4 | NP_008969.2 | 3 | 2030 | ||
GenBank mRNA | AF141293.1 | - | 1 | 65 | ||
AF142099.1 | - | 3 | 1423 | |||
AK094057.1 | - | 3 | 2029 | |||
BC093775.1 | - | 3 | 1385 | 7939620 | ||
BC093777.1 | - | 3 | 1385 | 7939622 |