Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001976.2 | NP_001001976.1 | 12 | 1552 | ||
NM_001288734.1 | NP_001275663.1 | 11 | 1448 | |||
NM_001288735.1 | NP_001275664.1 | 10 | 1425 | |||
NM_001288736.1 | NP_001275665.1 | 12 | 1787 | |||
NM_007041.3 | NP_008972.2 | 12 | 1552 | |||
NR_110006.1 | 10 | 1494 | ||||
XM_005269458.2 | XP_005269515.1 | 13 | 1693 | |||
XM_005269459.3 | XP_005269516.1 | 13 | 1916 | |||
XM_017015597.1 | XP_016871086.1 | 10 | 1362 | |||
XM_017015598.1 | XP_016871087.1 | 12 | 1771 | |||
XM_017015599.1 | XP_016871088.1 | 11 | 1435 | |||
XM_017015600.1 | XP_016871089.1 | 12 | 1415 | |||
XM_017015602.1 | XP_016871091.1 | 11 | 1332 | |||
XM_017015604.1 | XP_016871093.1 | 10 | 1277 | |||
GenBank mRNA | AF079098.1 | - | 12 | 1426 | ||
AF079099.1 | - | 12 | 1426 | |||
AK124669.1 | - | 10 | 1502 | |||
AK292657.1 | - | 7 | 668 | |||
AK295642.1 | - | 7 | 784 | |||
AK296356.1 | - | 11 | 1286 | |||
AK299101.1 | - | 12 | 1438 | |||
AK303604.1 | - | 10 | 1383 | |||
AK308926.1 | - | 5 | 581 | |||
AK313057.1 | - | 12 | 1512 | |||
BC022026.1 | - | 12 | 1525 | 4825006 |