Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206999.1 | NP_001193928.1 | 45 | 5717 | ||
NM_007174.2 | NP_009105.1 | 44 | 5591 | |||
XM_006719206.2 | XP_006719269.1 | 45 | 5699 | |||
XM_011537783.1 | XP_011536085.1 | 46 | 5789 | |||
XM_011537784.1 | XP_011536086.1 | 46 | 5789 | |||
XM_011537785.1 | XP_011536087.1 | 46 | 5744 | |||
XM_011537787.1 | XP_011536089.1 | 44 | 5573 | |||
XM_011537788.1 | XP_011536090.1 | 44 | 5573 | |||
XM_011537789.1 | XP_011536091.1 | 38 | 4924 | |||
XM_011537790.2 | XP_011536092.1 | 37 | 4492 | |||
XM_011537791.2 | XP_011536093.1 | 37 | 5477 | |||
XM_011537792.2 | XP_011536094.1 | 36 | 5532 | |||
XM_017018735.1 | XP_016874224.1 | 45 | 5744 | |||
XM_017018736.1 | XP_016874225.1 | 46 | 5744 | |||
XM_017018737.1 | XP_016874226.1 | 45 | 5699 | |||
XM_017018738.1 | XP_016874227.1 | 35 | 5033 | |||
XM_017018739.1 | XP_016874228.1 | 35 | 5808 | |||
GenBank mRNA | AB023166.2 | - | 36 | 5532 | ||
AY257469.1 | - | 44 | 5588 | |||
AY681966.1 | - | 45 | 5717 |