Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLASRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278439.1 | NP_001265368.1 | 3, 4 | 233 | ||
NM_007056.2 | NP_008987.2 | 5 | 419 | |||
NR_103529.1 | 5 | 419 | ||||
XM_011526396.2 | XP_011524698.1 | 5 | 414 | |||
XM_011526397.2 | XP_011524699.1 | 4 | 264 | |||
GenBank mRNA | AF042800.1 | - | 5 | 419 | ||
AK094681.1 | - | 4 | 1013 | |||
AK293333.1 | - | 3, 4 | 229 | |||
AY358944.1 | - | 5 | 400 | |||
BC013178.1 | - | 5 | 364 | |||
BC080554.1 | - | 4 | 359 | 6258057 | ||
BC131496.1 | - | 4 | 277 |