Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007059.3 | NP_008990.2 | 2 | 346 | ||
NR_111923.1 | 2 | 346 | ||||
XM_011526399.2 | XP_011524701.1 | 5 | 1851 | |||
XM_017026226.1 | XP_016881715.1 | 5 | 696 | |||
XM_017026227.1 | XP_016881716.1 | 4 | 772 | |||
XM_017026228.1 | XP_016881717.1 | 5 | 696 | |||
XM_017026229.1 | XP_016881718.1 | 5 | 696 | |||
GenBank mRNA | AF105369.1 | - | 2 | 278 | ||
AK023996.1 | - | 2 | 301 | |||
AK225419.1 | - | 2 | 290 | |||
AK298672.1 | - | 2 | 244 | |||
BC009249.2 | - | 2 | 261 | 3029623 | ||
DA129265.1 | - | 2 | 346 | |||
DA824599.1 | - | 2 | 350 |