Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126335.1 | NP_001119807.1 | 9 | 1025 | ||
NM_001243036.1 | NP_001229965.1 | 9 | 1120 | |||
NM_014270.4 | NP_055085.1 | 9 | 1111 | |||
XM_006722992.1 | XP_006723055.1 | 7 | 786 | |||
XM_011526402.2 | XP_011524704.1 | 9 | 2215 | |||
XM_017026230.1 | XP_016881719.1 | 7 | 761 | |||
GenBank mRNA | AB033548.1 | - | 9 | 996 | ||
AF141289.1 | - | 9 | 1093 | |||
AJ249199.1 | - | 8 | 970 | |||
AK026446.1 | - | 9 | 1094 | |||
AK223147.1 | - | 9 | 1099 | |||
AK313708.1 | - | 9 | 1012 | |||
AL365340.1 | - | 2 | 163 | |||
AY170373.1 | - | 5 | 539 | |||
BC017962.1 | - | 9 | 1100 | 4608486 |