Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126335.1 | NP_001119807.1 | 9, 10 | 1092 | ||
NM_001243036.1 | NP_001229965.1 | 9, 10 | 1187 | |||
NM_014270.4 | NP_055085.1 | 9, 10 | 1178 | |||
XM_006722992.1 | XP_006723055.1 | 7, 8 | 853 | |||
XM_011526402.2 | XP_011524704.1 | 9, 10 | 2282 | |||
XM_017026230.1 | XP_016881719.1 | 7, 8 | 828 | |||
GenBank mRNA | AB033548.1 | - | 9, 10 | 1063 | ||
AF141289.1 | - | 9, 10 | 1160 | |||
AJ249199.1 | - | 8, 9 | 1037 | |||
AK026446.1 | - | 9, 10 | 1161 | |||
AK223147.1 | - | 9, 10 | 1166 | |||
AK313708.1 | - | 9, 10 | 1079 | |||
AL365340.1 | - | 2, 3 | 230 | |||
AL365341.1 | - | 1, 2 | 14 | |||
BC017962.1 | - | 9, 10 | 1167 | 4608486 |