Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102426.1 | NP_001095896.1 | 11, 12 | 2033 | ||
XM_005263862.3 | XP_005263919.1 | 11, 12 | 2322 | |||
XM_005263863.4 | XP_005263920.1 | 11, 12 | 2109 | |||
XM_005263864.3 | XP_005263921.1 | 11, 12 | 2274 | |||
XM_011510507.1 | XP_011508809.1 | 13, 14 | 2164 | |||
XM_017003198.1 | XP_016858687.1 | 10, 11 | 2229 | |||
XM_017003199.1 | XP_016858688.1 | 10, 11 | 2182 | |||
GenBank mRNA | AB017114.1 | - | 7, 8 | 1161 | ||
AB449889.1 | - | 11, 12 | 1902 | |||
AK292826.1 | - | 9, 10 | 1666 | |||
BQ673509.1 | - | 2, 3 | 251 |