Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029896.1 | NP_001025067.1 | 11 | 1162 | ||
NM_007075.3 | NP_009006.2 | 12 | 1432 | |||
GenBank mRNA | AJ005897.1 | - | 11 | 1118 | ||
AK225122.1 | - | 11 | 1141 | |||
AK315717.1 | - | 11 | 1149 | |||
AY691428.1 | - | 10 | 990 | |||
BC000464.2 | - | 11 | 1156 | 2822742 | ||
BC003037.1 | - | 11 | 1156 | 2822742 | ||
BC009027.1 | - | 8 | 819 | |||
BC069206.1 | - | 12 | 1432 | 5801768 | ||
CR456725.1 | - | 10 | 990 |