Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321647.1 | NP_001308576.1 | 9 | 1004 | ||
NM_001321648.1 | NP_001308577.1 | 11 | 1108 | |||
NM_014142.3 | NP_054861.2 | 10 | 1013 | |||
GenBank mRNA | AK314144.1 | - | 10 | 999 | ||
BC000025.2 | - | 10 | 924 | 3505623 |
Gene Symbol: SEC61A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024576.2 | 11, 12 | 1400 | |||
NR_024577.2 | 12, 13 | 1533 | ||||
GenBank mRNA | AK057532.1 | - | 12, 13 | 1474 |