Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008396.2 | NP_001008397.1 | 9 | 854 | ||
NM_007086.3 | NP_009017.1 | 10 | 947 | |||
XM_006720012.2 | XP_006720075.1 | 10 | 945 | |||
XM_011536373.2 | XP_011534675.1 | 9 | 858 | |||
XM_017020940.1 | XP_016876429.1 | 9 | 858 | |||
XM_017020941.1 | XP_016876430.1 | 7 | 668 | |||
GenBank mRNA | AJ006266.1 | - | 9 | 896 | ||
AK293005.1 | - | 10 | 914 | |||
BC000622.2 | - | 10 | 912 | |||
BC005040.1 | - | 10 | 912 | |||
BC043349.1 | - | 10 | 935 | |||
BC063041.1 | - | 10 | 923 | 6514641 |