Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017585.3 | NP_060055.2 | 8 | 1144 | ||
XM_011518189.2 | XP_011516491.2 | 8 | 1121 | |||
XM_017014237.1 | XP_016869726.1 | 8 | 1112 | |||
XM_017014238.1 | XP_016869727.1 | 8 | 1116 | |||
GenBank mRNA | AB209108.1 | - | 4 | 3285 | ||
AJ011372.2 | - | 8 | 1086 | |||
AK074927.1 | - | 8 | 1127 | |||
AK096630.1 | - | 6 | 2021 | |||
AK222919.1 | - | 8 | 1089 | |||
AK225106.1 | - | 8 | 1017 | |||
BC013740.1 | - | 8 | 1078 | 4449594 | ||
Y17803.1 | - | 8 | 1095 |