Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP4K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042600.2 | NP_001036065.1 | 15, 16 | 1257 | ||
NM_007181.5 | NP_009112.1 | 15, 16 | 1257 | |||
XM_011526403.1 | XP_011524705.1 | 15, 16 | 1155 | |||
XM_011526404.1 | XP_011524706.1 | 15, 16 | 1155 | |||
XM_017026231.1 | XP_016881720.1 | 15, 16 | 1163 | |||
GenBank mRNA | AK303267.1 | - | 6, 7 | 487 | ||
U66464.1 | - | 15, 16 | 1171 |