Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NISCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276293.1 | NP_001263222.1 | 3 | 323 | ||
NM_001276294.1 | NP_001263223.1 | 3 | 323 | |||
NM_007184.3 | NP_009115.2 | 3 | 323 | |||
XM_005264839.2 | XP_005264896.1 | 2 | 96 | |||
GenBank mRNA | AB023192.1 | - | 4 | 261 | ||
AF082516.1 | - | 3 | 215 | |||
AK027428.1 | - | 3 | 201 | |||
AK297657.1 | - | 3 | 227 | |||
AK315001.1 | - | 3 | 227 | |||
AL703940.1 | - | 3 | 197 | |||
BC038102.2 | - | 3 | 227 | 5268247 | ||
BC054494.1 | - | 3 | 221 | 6195532 | ||
BC056900.1 | - | 3 | 193 | 6143316 | ||
DB496030.1 | - | 3 | 323 |