Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318219.1 | NP_001305148.1 | 20 | 3094 | ||
NM_007186.5 | NP_009117.2 | 22 | 3512 | |||
XM_005260262.4 | XP_005260319.1 | 22 | 3265 | |||
XM_005260263.4 | XP_005260320.1 | 22 | 3265 | |||
XM_005260264.4 | XP_005260321.1 | 21 | 3034 | |||
XM_006723690.3 | XP_006723753.1 | 22 | 3262 | |||
XM_006723691.1 | XP_006723754.1 | 22 | 3478 | |||
XM_006723692.3 | XP_006723755.1 | 21 | 3188 | |||
XM_006723693.3 | XP_006723756.1 | 21 | 3172 | |||
XM_006723694.3 | XP_006723757.1 | 22 | 3265 | |||
XM_011528517.2 | XP_011526819.1 | 22 | 3265 | |||
XM_011528518.2 | XP_011526820.1 | 22 | 3269 | |||
XM_011528519.2 | XP_011526821.1 | 15 | 2378 | |||
XM_017027617.1 | XP_016883106.1 | 22 | 3265 | |||
XM_017027618.1 | XP_016883107.1 | 14 | 2365 | |||
XM_017027619.1 | XP_016883108.1 | 11 | 1632 | |||
GenBank mRNA | AF022655.1 | - | 20 | 3086 | ||
AF049105.1 | - | 23 | 3536 | |||
BC144706.1 | - | 20 | 3040 |