Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007193.4 | NP_009124.2 | 9 | 829 | ||
XM_011531571.2 | XP_011529873.1 | 10 | 901 | |||
GenBank mRNA | AF196478.1 | - | 9 | 791 | ||
AY626137.1 | - | 4 | 328 | |||
BC007320.2 | - | 9 | 809 | 2988009 |