Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007197.3 | NP_009128.1 | 1 | 2143 | ||
GenBank mRNA | AB027464.1 | - | 1 | 1676 | ||
BC070037.1 | - | 1 | 2144 | 30337870 | ||
BC074997.2 | - | 1 | 1680 | 30915397 | ||
BC074998.2 | - | 1 | 1680 | 30915609 |
Set Membership: |
G-protein coupled receptor |